Search
Search

Atp13a3| Gene Symbol: Atp13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128094.1 | NP_001121566.1 | 31 - 32 | 3659 | 60 | |
| NM_001128096.1 | NP_001121568.1 | 31 - 32 | 3659 | 60 | ||
Set Membership: |
|