Search
Search

Lypd5| Gene Symbol: Lypd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029806.1 | NP_084082.1 | 2 - 3 | 209 | 69 | |
| GenBank mRNA | AK008654.1 | - | 2 - 3 | 212 | 69 | |
| BC107186.2 | - | 2 - 3 | 209 | 40050796 | 69 | |
| BC107187.2 | - | 2 - 3 | 209 | 40050800 | 69 | |
| BC119485.1 | - | 2 - 3 | 209 | 40050803 | 69 | |
| BC119486.1 | - | 2 - 3 | 209 | 40050804 | 69 | |
Set Membership: |
|