Search
Search

Clhc1| Gene Symbol: Clhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163521.1 | NP_001156993.1 | 4 - 5 | 779 | 145 | |
| NM_028504.1 | NP_082780.1 | 4 - 5 | 812 | 145 | ||
| GenBank mRNA | AK006591.1 | - | 4 - 5 | 813 | 145 | |
| BC137683.1 | - | 4 - 5 | 736 | 8860702 | 145 | |
| BC144828.1 | - | 4 - 5 | 703 | 9053376 | 145 | |
Set Membership: |
|