Search
Search

Olfr1330| Gene Symbol: Olfr1330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146334.2 | NP_666446.1 | 1 | 457 | 94 | |
| GenBank mRNA | BC108943.1 | - | 1 | 457 | 40042936 | 94 |
| BC108944.1 | - | 1 | 452 | 94 | ||
Set Membership: |
|