| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800003212.mm.1 |
NR_044988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003212.mm.1 |
NR_044988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003212.mm.1 |
TC0800000468.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800003211.mm.1 |
NR_044988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003211.mm.1 |
NR_044988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003211.mm.1 |
TC0800000468.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800003213.mm.1 |
NR_044988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003213.mm.1 |
NR_044988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800003213.mm.1 |
TC0800000468.mm.1 |
MTA TC |