Search
Search

A830010M20Rik| Gene Symbol: A830010M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007574.2 | NP_001007575.2 | 2 - 3 | 162 | 71 | |
| NM_001168557.1 | NP_001162029.1 | 2 - 3 | 128 | 71 | ||
| GenBank mRNA | AK043587.1 | - | 2 - 3 | 163 | 71 | |
| AK043720.1 | - | 2 - 3 | 132 | 71 | ||
| AK122444.1 | - | 2 - 3 | 111 | 71 | ||
| AK164388.1 | - | 2 - 3 | 132 | 71 | ||
| AK169666.1 | - | 2 - 3 | 129 | 71 | ||
Set Membership: |
|