Search
Search

Tmem130| Gene Symbol: Tmem130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177735.4 | NP_808403.2 | 3 - 4 | 588 | 74 | |
| GenBank mRNA | BC067004.1 | - | 3 - 4 | 588 | 6416222 | 74 |
| BC127616.1 | - | 3 - 4 | 565 | 40093666 | 74 | |
| BC127617.1 | - | 3 - 4 | 565 | 40093667 | 74 | |
Set Membership: |
|