| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500021747.mm.1 |
TC0500002410.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021747.mm.1 |
NM_001256059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021747.mm.1 |
NM_001256059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021748.mm.1 |
TC0500002410.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021748.mm.1 |
NM_001256059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021748.mm.1 |
NM_001256059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021746.mm.1 |
TC0500002410.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021746.mm.1 |
NM_001256059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021746.mm.1 |
NM_001256059.1 |
Refseq |