Search
Search

Phactr3| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007154.3 | NP_001007155.1 | 6 - 7 | 913 | 64 | |
| NM_001177789.1 | NP_001171260.1 | 6 - 7 | 1131 | 64 | ||
| NM_001177790.1 | NP_001171261.1 | 6 - 7 | 1131 | 64 | ||
| NM_001177791.1 | NP_001171262.1 | 6 - 7 | 965 | 64 | ||
| NM_028806.2 | NP_083082.1 | 6 - 7 | 1370 | 64 | ||
| GenBank mRNA | AJ632220.1 | - | 6 - 7 | 928 | 64 | |
| AJ632221.1 | - | 6 - 7 | 966 | 64 | ||
| AJ632222.1 | - | 6 - 7 | 913 | 64 | ||
| AJ632223.1 | - | 5 - 6 | 803 | 64 | ||
| AK005136.1 | - | 6 - 7 | 935 | 64 | ||
| AK032242.1 | - | 6 - 7 | 1137 | 64 | ||
| AK038449.1 | - | 6 - 7 | 881 | 64 | ||
| AK039213.1 | - | 6 - 7 | 920 | 64 | ||
| AK135144.1 | - | 6 - 7 | 1371 | 64 | ||
| AK220554.1 | - | 6 - 7 | 1131 | 64 | ||
| AY501387.1 | - | 6 - 7 | 1077 | 64 | ||
| BC058621.1 | - | 6 - 7 | 1286 | 5708186 | 64 | |
| BC059869.1 | - | 6 - 7 | 1149 | 6809310 | 64 | |
Set Membership: |
|