| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
461735 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
29497 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399727 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
460975 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
AK042363.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
AK049073.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
AK088621.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
BC100310.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
BC131929.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
BC131931.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363043 |
NM_177073.6 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536747 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384907 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
847408 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58227 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P103259 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P108468 |
AW494830.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P108468 |
BC049685.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P108468 |
AK012157.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P103259 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08086048 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02834941 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108468 |
AW494830.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108468 |
BC049685.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108468 |
AK012157.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P103259 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11766269 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11135150 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543347 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543333 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543336 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543346 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543348 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543338 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543337 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11766265 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11687972 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543334 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29543345 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11766267 |
- |
- |