Search
Search

Tmem214| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144525.3 | NP_653108.2 | 1 - 2 | 170 | 67 | |
| GenBank mRNA | AK034883.1 | - | 1 - 2 | 172 | 67 | |
| AK053521.1 | - | 186 | 67 | |||
| AK143078.1 | - | 172 | 67 | |||
| AK165285.1 | - | 1 - 2 | 182 | 67 | ||
| AK166510.1 | - | 1 - 2 | 165 | 67 | ||
| BC026651.1 | - | 1 - 2 | 86 | 4987771 | 67 | |
| BC029150.1 | - | 1 - 2 | 158 | 4954376 | 67 | |
Set Membership: |
|