Search
Search

4933413J09Rik| Gene Symbol: 4933413J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038005.1 | 2 - 3 | 225 | 121 | ||
| GenBank mRNA | AK016807.1 | - | 4 - 5 | 475 | 121 | |
| AK087762.1 | - | 2 - 3 | 226 | 121 | ||
| DV663125.1 | - | 244 | 121 | |||
| HM015003.1 | - | 3 - 4 | 446 | 121 | ||
Set Membership: |
|