| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016952 |
NR_027975.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016952 |
BC066076.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132859 |
NR_027975.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132859 |
BC066076.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
487128 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576106 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
8515 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697868 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187025 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899771 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333124 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
782218 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
238943 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174787 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
468869 |
AK040269.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831135 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9352 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
389834 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649578 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
343886 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
48367 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
125648 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969336 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
432663 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
37089 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593847 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546704 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851141 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63744 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535922 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277627 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
AF119498.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
AK086863.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
AK164029.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
AK206446.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
BC019440.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
BC092256.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549253 |
NM_013859.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408648 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
93276 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
19531 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
166765 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75595 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016952 |
NR_027975.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016952 |
BC066076.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132859 |
NR_027975.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132859 |
BC066076.1 |
mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661138 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07004562 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07024614 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07290776 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661123 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661145 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661146 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661128 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661126 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661104 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661118 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661113 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661162 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661157 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661125 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661140 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07222009 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06828244 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07290782 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661150 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661119 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07437518 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661129 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661114 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661156 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661110 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661127 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661153 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661108 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661121 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661105 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661155 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661158 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07160366 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661154 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661136 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25661134 |
- |
- |