Search
Search

Fam135a| Gene Symbol: Fam135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026604.4 | NP_080880.4 | 12 - 13 | 1517 | 65 | |
| GenBank mRNA | AK053786.1 | - | 12 - 13 | 1436 | 65 | |
| AK149303.1 | - | 12 - 13 | 1509 | 65 | ||
| BC070446.1 | - | 11 - 12 | 1199 | 30362784 | 65 | |
Set Membership: |
|