Search
Search

Dmrtc1a| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038616.2 | NP_001033705.2 | 2 - 3 | 330 | 88 | |
| NM_027591.3 | NP_081867.1 | 4 - 5 | 428 | 88 | ||
| NM_029378.2 | NP_083654.1 | 2 - 3 | 242 | 88 | ||
| GenBank mRNA | AK006548.1 | - | 2 - 3 | 345 | 88 | |
| AK014934.1 | - | 4 - 5 | 429 | 88 | ||
| BC147175.1 | - | 4 - 5 | 400 | 9056223 | 88 | |
| BC147176.1 | - | 4 - 5 | 400 | 9056216 | 88 | |
| DQ316772.1 | - | 4 - 5 | 384 | 88 | ||
| DQ316773.1 | - | 2 - 3 | 242 | 88 | ||
| DQ316774.1 | - | 2 - 3 | 242 | 88 | ||
Set Membership: |
|