| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200042688.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042688.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042688.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042694.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042694.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042694.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042695.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042695.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042695.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042686.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042686.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042686.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042692.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042692.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042692.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042696.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042696.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042696.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042690.mm.1 |
NR_040505.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042690.mm.1 |
NR_040505.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042690.mm.1 |
TC0200004997.mm.1 |
MTA TC |
| Murine Genome U74v2 Set |
168790_PM_at |
- |
- |
| Murine Genome U74v2 Set |
168790_at |
- |
- |