Search
Search

Pde4b| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177980.1 | NP_001171451.1 | 2 - 3 | 909 | 73 | |
| NM_001177981.1 | NP_001171452.1 | 2 - 3 | 449 | 73 | ||
| NM_001177982.1 | NP_001171453.1 | 2 - 3 | 185 | 73 | ||
| NM_019840.2 | NP_062814.2 | 7 - 8 | 1074 | 73 | ||
| GenBank mRNA | AF208023.2 | - | 7 - 8 | 1009 | 73 | |
| AF326555.1 | - | 7 - 8 | 814 | 73 | ||
| AF326556.1 | - | 7 - 8 | 814 | 73 | ||
| AJ297397.1 | - | 7 - 8 | 1009 | 73 | ||
| AK051102.1 | - | 2 - 3 | 450 | 73 | ||
| AK150868.1 | - | 2 - 3 | 415 | 73 | ||
| AK161341.1 | - | 2 - 3 | 186 | 73 | ||
| BB654856.1 | - | 2 - 3 | 450 | 73 | ||
| BC007155.1 | - | 2 - 3 | 256 | 73 | ||
| BC023751.1 | - | 2 - 3 | 885 | 73 | ||
| BC071259.1 | - | 2 - 3 | 425 | 5368075 | 73 | |
Set Membership: |
|