Search
Search

Trim9| Gene Symbol: Trim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110202.1 | NP_001103672.1 | 2 - 3 | 1364 | 59 | |
| NM_001110203.1 | NP_001103673.1 | 2 - 3 | 1364 | 59 | ||
| NM_001286386.1 | NP_001273315.1 | 2 - 3 | 1364 | 59 | ||
| NM_001286387.1 | NP_001273316.1 | 2 - 3 | 1364 | 59 | ||
| NM_001286388.1 | NP_001273317.1 | 2 - 3 | 1364 | 59 | ||
| NM_053167.3 | NP_444397.2 | 2 - 3 | 1364 | 59 | ||
| GenBank mRNA | AK028280.1 | - | 2 - 3 | 1083 | 59 | |
| AK035807.1 | - | 2 - 3 | 1082 | 59 | ||
| AK048875.1 | - | 2 - 3 | 1101 | 59 | ||
| AK049907.1 | - | 2 - 3 | 1078 | 59 | ||
| AK129109.1 | - | 2 - 3 | 1069 | 59 | ||
| AK163123.1 | - | 2 - 3 | 1101 | 59 | ||
| BC052034.1 | - | 2 - 3 | 1364 | 5684877 | 59 | |
Set Membership: |
|