Search
Search

Ccdc171| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081012.1 | NP_001074481.1 | 13 - 14 | 1827 | 85 | |
| GenBank mRNA | BC042767.1 | - | 1 - 2 | 70 | 85 | |
| BC172165.1 | - | 12 - 13 | 1618 | 85 | ||
Set Membership: |
|