| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10511068 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10511064 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10511067 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10511065 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10511063 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1460451_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10511062 |
- |
- |
| Mouse Genome 430 2.0 Array |
1460451_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1460451_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69671_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69671_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_001253856.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_001253856.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_001253858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_001253858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_027161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
NM_027161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017208.mm.1 |
TC0400002076.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_001253856.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_001253856.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_001253858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_001253858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_027161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
NM_027161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017209.mm.1 |
TC0400002076.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_001253856.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_001253856.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_001253858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_001253858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_027161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
NM_027161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017210.mm.1 |
TC0400002076.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_001253856.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_001253856.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_001253858.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_001253858.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_027161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
NM_027161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017207.mm.1 |
TC0400002076.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1460451_at |
- |
- |
| Mouse Expression Set 430 |
1460451_PM_at |
- |
- |
| Murine Genome U74v2 Set |
167219_PM_r_at |
- |
- |
| Murine Genome U74v2 Set |
129231_PM_at |
- |
- |
| Murine Genome U74v2 Set |
166990_f_at |
- |
- |
| Murine Genome U74v2 Set |
129231_at |
- |
- |
| Murine Genome U74v2 Set |
166990_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
167219_r_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1460451_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1460451_at |
- |
- |