| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152868 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152869 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152867 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152871 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1961938 |
AB087505.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
908972 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
88547 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658920 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572568 |
AK005793.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572568 |
AK047371.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572568 |
AK047938.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572568 |
XM_001473444.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
820536 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692927 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
949977 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
99167 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151076 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66735 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P107954 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139892 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1961938 |
AB087505.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152858 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152867 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152888 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152860 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152861 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152879 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152874 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152864 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152876 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152869 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152886 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152866 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152871 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152857 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152885 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152883 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152881 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152882 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28152877 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P107954 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139892 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P238665 |
AB087505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238665 |
AK009293.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238665 |
AK043266.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238665 |
NM_027132.2 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P107954 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139892 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09085876 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09085893 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09728819 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09085905 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152868 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152871 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152869 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28152867 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152865 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152867 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152921 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152876 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152953 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152886 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152814 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152916 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152883 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153011 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152950 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152810 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152908 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153008 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152871 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152910 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152917 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152864 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152877 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152861 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28152892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28153020 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152879 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09500858 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152950 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152869 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152916 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152921 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09500865 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152877 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152956 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152910 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152952 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152876 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152865 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152861 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152862 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152885 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152917 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152888 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09530540 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152895 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09085874 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152871 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152951 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152866 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152898 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152882 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152900 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152920 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152903 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152858 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152867 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152889 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09728819 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152881 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09085886 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152908 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152897 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09085904 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152915 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152905 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152953 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09673592 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28152874 |
- |
- |