| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665423 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665422 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665420 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1988970 |
BC080765.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1988970 |
BC100336.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1988970 |
AK004987.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1988970 |
NM_021527.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1988970 |
NM_001141946.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489325 |
AK076650.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489325 |
NM_172895.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402951 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45154 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702811 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788418 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242607 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
64477 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859070 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
239779 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
591296 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
513919 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
163753 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113531 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110284 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114325 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1988970 |
BC080765.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1988970 |
BC100336.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1988970 |
AK004987.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1988970 |
NM_021527.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1988970 |
NM_001141946.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121288 |
BC117835.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121288 |
BC117836.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121288 |
BC080765.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121288 |
BC100336.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121288 |
AK032528.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665390 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665415 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665393 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665410 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665422 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665399 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665400 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665413 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665396 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665391 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665409 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665397 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665417 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665403 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665401 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665411 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665406 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665394 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21665419 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110284 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00503894 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114325 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04707246 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P541939 |
BY735964.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P541939 |
BC080765.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P541939 |
BC100336.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P541939 |
BC058174.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P541939 |
AK032528.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P121288 |
BC117835.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P121288 |
BC117836.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P121288 |
BC080765.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P121288 |
BC100336.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P121288 |
AK032528.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129288 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114325 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110284 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01487875 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01487873 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02227834 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665422 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665420 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21665423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665379 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665396 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665406 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665322 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665345 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665337 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665419 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665347 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665356 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665324 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665386 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665371 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665343 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665339 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665305 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665355 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665415 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665353 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665367 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665323 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665393 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665349 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665341 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665420 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665410 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665336 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665308 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665357 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665409 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665318 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665312 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665320 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665399 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665315 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665376 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665401 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665422 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665361 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665307 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665394 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21665333 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665394 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665386 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665356 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01999722 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665345 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665388 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665305 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01801530 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665379 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01147479 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01231954 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01277982 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665310 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665378 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01801529 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665308 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665351 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665307 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665376 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665422 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01868341 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665353 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665391 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665370 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665393 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665358 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665384 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665303 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665349 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665341 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665420 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665383 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665415 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665405 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01277987 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665417 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665406 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665400 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01485363 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665402 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01277991 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665339 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665304 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665413 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665409 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665397 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665382 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01974179 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665396 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665374 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01974176 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665306 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665355 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665401 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665343 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665419 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665347 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21665399 |
- |
- |