Search
Search

Gm8801| Gene Symbol: Gm8801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028278.1 | 2 - 3 | 461 | 80 | ||
| GenBank mRNA | CA558512.1 | - | 2 - 3 | 461 | 80 | |
| CF916400.1 | - | 2 - 3 | 404 | 80 | ||
| DV649894.1 | - | 2 - 3 | 439 | 80 | ||
Set Membership: |
|