Search
Search
| Gene Symbol: Cd44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039150.1 | NP_001034239.1 | 2 - 3 | 565 | 83 | |
| NM_001039151.1 | NP_001034240.1 | 2 - 3 | 565 | 83 | ||
| NM_001177785.1 | NP_001171256.1 | 2 - 3 | 565 | 83 | ||
| NM_001177786.1 | NP_001171257.1 | 2 - 3 | 565 | 83 | ||
| NM_001177787.1 | NP_001171258.1 | 2 - 3 | 565 | 83 | ||
| NM_009851.2 | NP_033981.2 | 2 - 3 | 565 | 83 | ||
| GenBank mRNA | AJ251594.1 | - | 2 - 3 | 352 | 83 | |
| AK045226.1 | - | 2 - 3 | 411 | 83 | ||
| AK087472.1 | - | 2 - 3 | 359 | 83 | ||
| AK144125.1 | - | 2 - 3 | 368 | 83 | ||
| AK152099.1 | - | 2 - 3 | 361 | 83 | ||
| AK154411.1 | - | 2 - 3 | 356 | 83 | ||
| AK162720.1 | - | 2 - 3 | 361 | 83 | ||
| AK166320.1 | - | 2 - 3 | 350 | 83 | ||
| BC005676.1 | - | 2 - 3 | 344 | 83 | ||
| BC051388.1 | - | 2 - 3 | 395 | 4910789 | 83 | |
| CT010324.1 | - | 2 - 3 | 246 | 83 | ||
| J05163.1 | - | 2 - 3 | 227 | 83 | ||
| M27129.1 | - | 2 - 3 | 272 | 83 | ||
| M27130.1 | - | 2 - 3 | 278 | 83 | ||
| M30655.1 | - | 2 - 3 | 346 | 83 | ||
| X66081.1 | - | 2 - 3 | 249 | 83 | ||
| X66082.1 | - | 2 - 3 | 249 | 83 | ||
| X66083.1 | - | 2 - 3 | 249 | 83 | ||
| X66084.1 | - | 2 - 3 | 249 | 83 | ||
Set Membership: |
|