Search
Search

Klf11| Gene Symbol: Klf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178357.3 | NP_848134.1 | 3 - 4 | 1381 | 65 | |
| GenBank mRNA | AJ459773.2 | - | 3 - 4 | 1227 | 65 | |
| AK036618.1 | - | 3 - 4 | 1367 | 65 | ||
| AK038397.1 | - | 3 - 4 | 1479 | 65 | ||
| AK050156.1 | - | 3 - 4 | 1383 | 65 | ||
| AK054170.1 | - | 3 - 4 | 1353 | 65 | ||
| AK082838.1 | - | 3 - 4 | 1370 | 65 | ||
| AK084913.1 | - | 3 - 4 | 1354 | 65 | ||
| AK085319.1 | - | 3 - 4 | 1356 | 65 | ||
| AK089044.1 | - | 3 - 4 | 1361 | 65 | ||
| AK161233.1 | - | 3 - 4 | 1373 | 65 | ||
| BC060642.1 | - | 3 - 4 | 1379 | 6850350 | 65 | |
Set Membership: |
|