Search
Search
| Gene Symbol: Il19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009940.1 | NP_001009940.1 | 2 - 3 | 206 | 58 | |
| GenBank mRNA | AF453945.1 | - | 2 - 3 | 206 | 58 | |
| BC115793.1 | - | 3 - 4 | 265 | 40100999 | 58 | |
Set Membership: |
|