| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011301.mm.1 |
NM_177756.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011301.mm.1 |
NM_177756.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011301.mm.1 |
TC0100001365.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011302.mm.1 |
NM_177756.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011302.mm.1 |
NM_177756.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011302.mm.1 |
TC0100001365.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011304.mm.1 |
NM_177756.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011304.mm.1 |
NM_177756.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011304.mm.1 |
TC0100001365.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011303.mm.1 |
NM_177756.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011303.mm.1 |
NM_177756.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011303.mm.1 |
TC0100001365.mm.1 |
MTA TC |