Search
Search

Gm4846| Gene Symbol: Gm4846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164306.1 | NP_001157778.1 | 5 - 6 | 899 | 67 | |
| GenBank mRNA | BC147800.1 | - | 5 - 6 | 899 | 9087517 | 67 |
| BC147804.1 | - | 5 - 6 | 907 | 9087521 | 67 | |
Set Membership: |
|