| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
AF159571.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
AK144199.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
AK159242.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
AK160927.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
BC067248.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786761 |
NM_011819.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
499108 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
104047 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
184437 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671488 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323461 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
145567 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P178171 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P178171 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
BC022155.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
NM_001144993.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
AK013367.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P310164 |
NM_001144992.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115142 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P178171 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409958 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409946 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409985 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409927 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409925 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409932 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409981 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409971 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00316769 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409920 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409954 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409979 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01065403 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409990 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409928 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409931 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409978 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409984 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00529927 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409982 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409943 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01065408 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00529928 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00129058 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409969 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409970 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00604203 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409987 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409937 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409967 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409955 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409922 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409975 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00711853 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409938 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409934 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409966 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409919 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409923 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409976 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409989 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409960 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20409956 |
- |
- |