Search
Search

Prkaa1| Gene Symbol: Prkaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013367.3 | NP_001013385.3 | 6 - 7 | 827 | 65 | |
| GenBank mRNA | AK147811.1 | - | 6 - 7 | 804 | 65 | |
| AK160612.1 | - | 6 - 7 | 808 | 65 | ||
| AY885266.1 | - | 6 - 7 | 786 | 65 | ||
| BC086695.1 | - | 6 - 7 | 797 | 30691866 | 65 | |
| CB245934.1 | - | 1 - 2 | 123 | 65 | ||
Set Membership: |
|