Search
Search

Syngr1| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009303.2 | NP_033329.1 | 1 - 2 | 155 | 58 | |
| NM_207708.1 | NP_997591.1 | 1 - 2 | 155 | 58 | ||
| GenBank mRNA | AJ002306.1 | - | 1 - 2 | 135 | 58 | |
| AK002972.1 | - | 1 - 2 | 137 | 58 | ||
| AK010442.1 | - | 1 - 2 | 122 | 58 | ||
| AK044007.1 | - | 1 - 2 | 146 | 58 | ||
| AK153096.1 | - | 1 - 2 | 122 | 58 | ||
| AK153187.1 | - | 1 - 2 | 122 | 58 | ||
| AK156134.1 | - | 1 - 2 | 139 | 58 | ||
| AK194549.1 | - | 1 - 2 | 63 | 58 | ||
| BC048172.1 | - | 1 - 2 | 112 | 58 | ||
| BC138728.1 | - | 1 - 2 | 182 | 8861750 | 58 | |
| BC138729.1 | - | 1 - 2 | 193 | 8861751 | 58 | |
Set Membership: |
|