Search
Search

Pip4k2b| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054051.1 | NP_473392.1 | 2 - 3 | 356 | 77 | |
| GenBank mRNA | AK089414.1 | - | 2 - 3 | 346 | 77 | |
| AK146559.1 | - | 2 - 3 | 406 | 77 | ||
| AK170392.1 | - | 2 - 3 | 283 | 77 | ||
| AK171497.1 | - | 2 - 3 | 346 | 77 | ||
| AY050219.1 | - | 2 - 3 | 185 | 77 | ||
| BC047282.1 | - | 2 - 3 | 356 | 6491155 | 77 | |
Set Membership: |
|