| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100009316.mm.1 |
NM_177604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009316.mm.1 |
NM_177604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009316.mm.1 |
TC0100001161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009319.mm.1 |
NM_177604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009319.mm.1 |
NM_177604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009319.mm.1 |
TC0100001161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009317.mm.1 |
NM_177604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009317.mm.1 |
NM_177604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009317.mm.1 |
TC0100001161.mm.1 |
MTA TC |