Search
Search

2410089E03Rik| Gene Symbol: 2410089E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162906.1 | NP_001156378.1 | 15 - 16 | 3087 | 91 | |
| GenBank mRNA | AK028897.1 | - | 15 - 16 | 3088 | 91 | |
Set Membership: |
|