Search
Search

1500009C09Rik| Gene Symbol: 1500009C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037698.1 | 1 - 2 | 363 | 72 | ||
| GenBank mRNA | AK005178.1 | - | 1 - 2 | 265 | 72 | |
| BC023061.1 | - | 1 - 2 | 255 | 72 | ||
| BC094356.1 | - | 1 - 2 | 201 | 72 | ||
| BY281201.1 | - | 1 - 2 | 363 | 72 | ||
Set Membership: |
|