| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
615579 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
190286 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
405514 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505985 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
89258 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641243 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574068 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837883 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351606 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341261 |
AK089695.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
927903 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399091 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
251812 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865301 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
223280 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760758 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
AK014485.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
AK122398.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
AY550908.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC034075.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC047348.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC049162.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC049224.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC057458.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC057923.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
BC064751.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
NM_001081359.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665056 |
NM_001112721.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746417 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49645 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
706610 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
215877 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467764 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508837 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342342 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448462 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
143196 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10175 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
74928 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114349 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P119811 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P119811 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06241079 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06241053 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114349 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01548149 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01548169 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P677891 |
BC034069.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P677891 |
BC110691.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P677891 |
NM_001162532.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136227 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P119811 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06241079 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114349 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230840 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06315925 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05999157 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230844 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230673 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06033482 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230676 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06076528 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230865 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06076524 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230769 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230855 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06615378 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230876 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230849 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06319784 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230867 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230671 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05999155 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230853 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230680 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230839 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06033479 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230850 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230679 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230678 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230842 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230772 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230681 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230869 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230862 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25230878 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06319783 |
- |
- |