| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400004256.mm.1 |
NR_045959.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004256.mm.1 |
NR_045959.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004256.mm.1 |
TC1400000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004252.mm.1 |
NR_045959.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004252.mm.1 |
NR_045959.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004252.mm.1 |
TC1400000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004253.mm.1 |
NR_045959.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004253.mm.1 |
NR_045959.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004253.mm.1 |
TC1400000521.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400004254.mm.1 |
NR_045959.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004254.mm.1 |
NR_045959.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400004254.mm.1 |
TC1400000521.mm.1 |
MTA TC |