| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300024218.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024218.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024218.mm.1 |
TC0300003080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024220.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024220.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024220.mm.1 |
TC0300003080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024223.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024223.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024223.mm.1 |
TC0300003080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024219.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024219.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024219.mm.1 |
TC0300003080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024222.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024222.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024222.mm.1 |
TC0300003080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024221.mm.1 |
NM_139148.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024221.mm.1 |
NM_139148.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024221.mm.1 |
TC0300003080.mm.1 |
MTA TC |