Search
Search
| Gene Symbol: Rarb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289760.1 | NP_001276689.1 | 6 - 7 | 1190 | 72 | |
| NM_001289761.1 | NP_001276690.1 | 5 - 6 | 1109 | 72 | ||
| NM_001289762.1 | NP_001276691.1 | 4 - 5 | 719 | 72 | ||
| NM_011243.2 | NP_035373.1 | 4 - 5 | 1073 | 72 | ||
| GenBank mRNA | AK043726.1 | - | 4 - 5 | 1427 | 72 | |
| AK078471.1 | - | 4 - 5 | 785 | 72 | ||
| BC076597.1 | - | 4 - 5 | 1069 | 30608242 | 72 | |
| S56660.1 | - | 4 - 5 | 969 | 72 | ||
Set Membership: |
|