| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100030163.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030163.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030163.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030170.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030170.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030170.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030159.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030159.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030159.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030161.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030161.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030161.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030162.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030162.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030162.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030164.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030164.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030164.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030165.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030165.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030165.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030169.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030169.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030169.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030171.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030171.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030171.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030160.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030160.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030160.mm.1 |
TC0100003666.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030172.mm.1 |
NM_001081227.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030172.mm.1 |
NM_001081227.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030172.mm.1 |
TC0100003666.mm.1 |
MTA TC |