Search
Search

Becn2| Gene Symbol: Becn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290692.1 | NP_001277621.1 | 1 | 1322 | 59 | |
| GenBank mRNA | CJ207428.1 | - | 1 | 62 | 59 | |
| KF511582.1 | - | 1 | 1281 | 59 | ||
Set Membership: |
|