Search
Search

F13a1| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166391.1 | NP_001159863.1 | 10 - 11 | 1376 | 73 | |
| NM_028784.3 | NP_083060.2 | 10 - 11 | 1401 | 73 | ||
| GenBank mRNA | AK030914.1 | - | 11 - 12 | 1376 | 73 | |
| AK036403.1 | - | 10 - 11 | 1402 | 73 | ||
| AK049092.1 | - | 10 - 11 | 1402 | 73 | ||
| AK165311.1 | - | 10 - 11 | 1402 | 73 | ||
| BC011073.1 | - | 4 - 5 | 482 | 73 | ||
| BC040274.1 | - | 10 - 11 | 1392 | 3482233 | 73 | |
Set Membership: |
|