Search
Search

Leng8| Gene Symbol: Leng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172736.3 | NP_766324.1 | 5 - 6 | 587 | 62 | |
| GenBank mRNA | AK034310.1 | - | 5 - 6 | 592 | 62 | |
| AK170629.1 | - | 5 - 6 | 595 | 62 | ||
| AK200008.1 | - | 2 - 3 | 224 | 62 | ||
| BC041775.1 | - | 5 - 6 | 559 | 62 | ||
| BC042658.1 | - | 5 - 6 | 575 | 5365041 | 62 | |
| BC066768.1 | - | 5 - 6 | 578 | 6407146 | 62 | |
| CF536188.1 | - | 5 - 6 | 587 | 62 | ||
Set Membership: |
|