Search
Search

1700001O22Rik| Gene Symbol: 1700001O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198000.3 | NP_932117.2 | 4 - 5 | 894 | 69 | |
| NR_028486.2 | 4 - 5 | 411 | 69 | |||
| NR_028487.2 | 4 - 5 | 411 | 69 | |||
| GenBank mRNA | AK005618.1 | - | 4 - 5 | 405 | 69 | |
| AK076928.1 | - | 4 - 5 | 395 | 69 | ||
| BC023044.1 | - | 4 - 5 | 864 | 5355351 | 69 | |
| CD773886.1 | - | 3 - 4 | 69 | |||
Set Membership: |
|