Search
Search

Ptrh1| Gene Symbol: Ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178595.3 | NP_848710.1 | 3 - 4 | 394 | 67 | |
| GenBank mRNA | AK075833.1 | - | 3 - 4 | 395 | 67 | |
| BC098218.1 | - | 3 - 4 | 434 | 3466952 | 67 | |
Set Membership: |
|