Search
Search

2700094K13Rik| Gene Symbol: 2700094K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033166.2 | NP_001028338.1 | 1 - 2 | 225 | 53 | |
| NM_001037279.1 | NP_001032356.1 | 1 - 2 | 225 | 53 | ||
| GenBank mRNA | AK012609.1 | - | 1 - 2 | 203 | 53 | |
| AK142622.1 | - | 1 - 2 | 215 | 53 | ||
| AK167638.1 | - | 1 - 2 | 197 | 53 | ||
| AK171972.1 | - | 1 - 2 | 216 | 53 | ||
| BC056177.1 | - | 1 - 2 | 218 | 5683334 | 53 | |
| BY239096.1 | - | 1 - 2 | 287 | 53 | ||
| CA318611.1 | - | 1 - 2 | 122 | 53 | ||
Set Membership: |
|