Search
Search

Kif26a| Gene Symbol: Kif26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097621.1 | NP_001091090.1 | 12 - 13 | 5248 | 78 | |
| GenBank mRNA | AK129315.1 | - | 4 - 5 | 3375 | 78 | |
| BC059062.1 | - | 1 - 2 | 2079 | 78 | ||
| BC094358.1 | - | 1 - 2 | 2753 | 78 | ||
| GQ489027.1 | - | 12 - 13 | 5091 | 78 | ||
Set Membership: |
|