Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10608668 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10505248 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10505251 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10505260 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608668 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0400004725.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004725.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004725.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004726.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004726.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004726.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004727.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004727.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004727.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004724.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004724.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004724.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004723.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004723.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004723.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004728.mm.1 |
NM_001081285.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004728.mm.1 |
NM_001081285.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004728.mm.1 |
TC0400000663.mm.1 |
MTA TC |
Murine Genome U74v2 Set |
102096_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
102096_f_at |
- |
- |
Murine Genome U74v2 Set |
137167_at |
- |
- |
Murine Genome U74v2 Set |
137167_PM_at |
- |
- |