| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800004686.mm.1 |
NR_038181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004686.mm.1 |
NR_038181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004686.mm.1 |
NR_038189.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004686.mm.1 |
NR_038189.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004686.mm.1 |
TC0800000701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800004687.mm.1 |
NR_038189.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004687.mm.1 |
NR_038189.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004687.mm.1 |
TC0800000701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800004682.mm.1 |
NR_038181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004682.mm.1 |
NR_038181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004682.mm.1 |
NR_038189.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004682.mm.1 |
NR_038189.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004682.mm.1 |
TC0800000701.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800004683.mm.1 |
NR_038181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004683.mm.1 |
NR_038181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004683.mm.1 |
NR_038189.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004683.mm.1 |
NR_038189.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800004683.mm.1 |
TC0800000701.mm.1 |
MTA TC |