Search
Search
| Gene Symbol: Lsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015816.4 | NP_056631.2 | 2 - 3 | 270 | 85 | |
| GenBank mRNA | AJ249439.1 | - | 2 - 3 | 110 | 85 | |
| AK003847.1 | - | 2 - 3 | 106 | 85 | ||
| AK010910.1 | - | 2 - 3 | 106 | 85 | ||
| AK089028.1 | - | 2 - 3 | 123 | 85 | ||
| AK143470.1 | - | 2 - 3 | 94 | 85 | ||
| AK167784.1 | - | 2 - 3 | 134 | 85 | ||
| BC026747.1 | - | 2 - 3 | 253 | 4024321 | 85 | |
| CA322716.1 | - | 2 - 3 | 270 | 85 | ||
Set Membership: |
|